Merck
Clobazam therapeutic drug monitoring: a comprehensive review of the literature with proposals to improve future studies.
Therapeutic drug monitoring (2013-01-16)
Jose de Leon, Edoardo Spina, Francisco J Diaz
ABSTRAKT
Clobazam was recently approved for Lennox-Gastaut syndrome in the United States. There is no published review article focused on clobazam therapeutic drug monitoring (TDM) in English. More than 200 clobazam articles identified by a PubMed search were carefully reviewed for information on clobazam pharmacokinetics. Clobazam is mainly metabolized by a cytochrome P450 (CYP) isoenzyme, CYP3A4, to its active metabolite, N-desmethylclobazam. Then, N-desmethylclobazam is mainly metabolized by CYP2C19 unless the individual has no CYP2C19 activity [poor metabolizer (PM)]. Using a mechanistic approach to reinterpret the published findings of steady-state TDM and single-dosing pharmacokinetic studies, 4 different serum clobazam concentration ratios were studied. The available limited steady-state TDM data suggest that the serum N-desmethylclobazam/clobazam ratio can be useful for clinicians, including identifying CYP2C19 PMs (ratio >25 in the absence of inhibitors). There are 3 possible concentration/dose (C/D) ratios. The clobazam C/D ratio has the potential to measure the contribution of CYP3A4 activity to the clearance of clobazam from the body. The N-desmethylclobazam C/D ratio does not seem to be a good measure of clobazam clearance and should be substituted with the total (clobazam + N-desmethylclobazam) C/D ratio. Future clobazam TDM studies need to use trough concentrations after steady state has been reached (>3 weeks in normal individuals and several months in CYP2C19 PMs). These future studies need to explore the potential of clobazam and total C/D ratios. Better studies on the relative potency of N-desmethylclobazam compared with the parent compound are needed to provide weighted total serum concentrations that correct for the possible lower N-desmethylclobazam pharmacodynamic activity. Standardization and more studies of C/D ratios from clobazam and other drugs can be helpful to move TDM forward.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Clobazam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®