Merck
Progress in the studies on tryptanthrin, an alkaloid of history.
Archives of pharmacal research (2013-04-02)
Yurngdong Jahng
ABSTRAKT
Tryptanthrin, an indoloquinazoline alkaloid, was first obtained by sublimation of natural indigo and later isolated from the culture of fungus Candida lipolytica and a variety of other natural sources. Tryptanthrin showed a variety of intriguing biological properties such as antibacterial, antifungal, antiprotozoal, and antiparasitic activities, inhibitory activities against COX-2, 5-LOX, NO synthase and PGE(2) expression, as well as cytotoxic and antitumor activities. Present review covers recent studies on the natural sources, biological activities and mechanisms of their actions, synthesis, structure-activity relationship, and metabolism of tryptanthrin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)