Merck
  • Can azathioprine 6-mercaptopurine treatment be withdrawn in patients with response in ulcerative colitis: what is the most appropriate time for this?
Can azathioprine 6-mercaptopurine treatment be withdrawn in patients with response in ulcerative colitis: what is the most appropriate time for this?
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology (2012-01-01)
Gökhan Kabaçam, Murat Törüner
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Azathioprine, ≥98%
Azathioprine, European Pharmacopoeia (EP) Reference Standard
Supelco
Azathioprine, Pharmaceutical Secondary Standard; Certified Reference Material