Merck
Bisphenol A and human health: a review of the literature.
Reproductive toxicology (Elmsford, N.Y.) (2013-09-03)
Johanna R Rochester
ABSTRAKT
There is growing evidence that bisphenol A (BPA) may adversely affect humans. BPA is an endocrine disruptor that has been shown to be harmful in laboratory animal studies. Until recently, there were relatively few epidemiological studies examining the relationship between BPA and health effects in humans. However, in the last year, the number of these studies has more than doubled. A comprehensive literature search found 91 studies linking BPA to human health; 53 published within the last year. This review outlines this body of literature, showing associations between BPA exposure and adverse perinatal, childhood, and adult health outcomes, including reproductive and developmental effects, metabolic disease, and other health effects. These studies encompass both prenatal and postnatal exposures, and include several study designs and population types. While it is difficult to make causal links with epidemiological studies, the growing human literature correlating environmental BPA exposure to adverse effects in humans, along with laboratory studies in many species including primates, provides increasing support that environmental BPA exposure can be harmful to humans, especially in regards to behavioral and other effects in children.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bisphenol A, ≥99%