Merck
Magnesium in depression.
Pharmacological reports : PR (2013-08-21)
Anna Serefko, Aleksandra Szopa, Piotr Wlaź, Gabriel Nowak, Maria Radziwoń-Zaleska, Michał Skalski, Ewa Poleszak
ABSTRAKT
Magnesium is one of the most essential mineral in the human body, connected with brain biochemistry and the fluidity of neuronal membrane. A variety of neuromuscular and psychiatric symptoms, including different types of depression, was observed in magnesium deficiency. Plasma/serum magnesium levels do not seem to be the appropriate indicators of depressive disorders, since ambiguous outcomes, depending on the study, were obtained. The emergence of a new approach to magnesium compounds in medical practice has been seen. Apart from being administered as components of dietary supplements, they are also perceived as the effective agents in treatment of migraine, alcoholism, asthma, heart diseases, arrhythmias, renal calcium stones, premenstrual tension syndrome etc. Magnesium preparations have an essential place in homeopathy as a remedy for a range of mental health problems. Mechanisms of antidepressant action of magnesium are not fully understood yet. Most probably, magnesium influences several systems associated with development of depression. The first information on the beneficial effect of magnesium sulfate given hypodermically to patients with agitated depression was published almost 100 years ago. Numerous pre-clinical and clinical studies confirmed the initial observations as well as demonstrated the beneficial safety profile of magnesium supplementation. Thus, magnesium preparations seem to be a valuable addition to the pharmacological armamentarium for management of depression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%