Merck
  • [Synthetic studies on natural products with aromatic nitrogen heterocycles based on development of the methods for the formation of aryl carbon-nitrogen bond].
[Synthetic studies on natural products with aromatic nitrogen heterocycles based on development of the methods for the formation of aryl carbon-nitrogen bond].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (2013-10-04)
Kentaro Okano
ABSTRAKT
Total syntheses of yatakemycin, PDE-II, dictyodendrins, and heptaphylline are described. This article focuses on the formation of aryl carbon-nitrogen bonds by two methods: first by an aromatic amination reaction using a combination of CuI and CsOAc, and then by a benzyne-mediated one-pot cyclization-functionalization sequence. The aryl amination reaction shows a high functional group compatibility and applicability for highly hindered substrates. The utility of this reaction has been fully demonstrated through a total synthesis of yatakemycin, which features the regioselective ring opening of (S)-epichlorohydrin with 2,6-dibromophenyllithium species and the efficient deprotection of benzyl groups of aryl benzyl ether with BCl3 in the presence of pentamethylbenzene. The copper-mediated aryl amination reaction was applied to a one-pot double aryl amination for facile access to the highly substituted pyrroloindole skeleton, which led to a concise total synthesis of PDE-II. A highly efficient total synthesis of dictyodendrins A-E was accomplished by the development of a novel benzyne-mediated one-pot indoline formation/cross-coupling sequence for the construction of a highly substituted key indoline intermediate. Peripheral substituents were introduced onto this intermediate in a modular fashion to complete the total synthesis of dictyodendrins A-E. The utility of the one-pot benzyne-mediated cyclization-functionalization reaction was demonstrated by the total synthesis of carbazole alkaloid, heptaphylline.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Węgiel aktywowany, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Węgiel - Szklisty, rod, 200mm, diameter 1.0mm, glassy carbon
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis