Merck
1-nitropyrene.
IARC monographs on the evaluation of carcinogenic risks to humans (1989-01-01)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Nitropyrene, ≥95%
1-Nitropyrene, BCR®, certified reference material