Merck
[Hypoglycemia induced or facilitated by disopyramide].
La Revue de medecine interne (1988-11-01)
C Seriès
MATERIAŁY
Numer produktu
Marka
Opis produktu
Disopyramide, European Pharmacopoeia (EP) Reference Standard