Merck
[Phenacetin abuse I. Occurrence, per capita consumption and costs of treatment].
Schweizerische medizinische Wochenschrift (1980-01-28)
M J Mihatsch, H O Hofer, F Gutzwiler, F P Brunner, H U Zollinger
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Standardowa temperatura topnienia fenacetyny, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)