Merck
Kepone: a literature summary.
Journal of environmental pathology and toxicology (1978-03-01)
J E Huff, H B Gerstner
PMID82598
ABSTRAKT
This overview provides a narrative survey of the reference literature on Kepone. The circumstances surrounding the "Hopewell incident" have increased the awareness and vigilance of state and federal agencies regarding human and environmental exposure to toxic chemicals. Highlights of this overview include history, nomenclature, uses, toxicology, environmental aspects, epidemiology, and conclusions. A chronologic history of Kepone is detailed in the Appendix.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chlordecone, PESTANAL®, analytical standard
Supelco
Chlordecone, 99%, vial of 1 g, (Kepone equiv.), Cerilliant®