Merck
[Primidone].
Nihon rinsho. Japanese journal of clinical medicine (1995-02-01)
K Furuno, H Kawasaki, Y Gomita
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Primidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Primidone, analytical standard
Primidone, European Pharmacopoeia (EP) Reference Standard