Merck
Antitumorigenic effects of limonene and perillyl alcohol against pancreatic and breast cancer.
Advances in experimental medicine and biology (1996-01-01)
P L Crowell, A Siar Ayoubi, Y D Burke
ABSTRAKT
Perillyl alcohol is a natural product from cherries and other edible plants. Perillyl alcohol and d-limonene, a closely related dietary monoterpene, have chemotherapeutic activity against pancreatic, mammary, and prostatic tumors. In addition, perillyl alcohol, limonene, and other dietary monoterpenes have chemopreventive activity. Several mechanisms may account for the antitumorigenic effects of monoterpenes. For example, many monoterpenes inhibit the post-translational isoprenylation of cell growth-regulatory proteins such as Ras. Perillyl alcohol induces apoptosis without affecting the rate of DNA synthesis in both liver and pancreatic tumor cells. In addition, monoterpene-treated, regressing rat mammary tumors exhibit increased expression of transforming growth factor beta concomitant with tumor remodeling/redifferentiation to a more benign phenotype. Monoterpenes are effective, nontoxic dietary antitumor agents which act through a variety of mechanisms of action and hold promise as a novel class of antitumor drugs for human cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-Perillyl alcohol, analytical standard
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, ≥95%, FG
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, 96%