Merck
  • Analysis of lysergic acid diethylamide: comparison of capillary electrophoresis with laser-induced fluorescence (CE-LIF) with conventional techniques.
Analysis of lysergic acid diethylamide: comparison of capillary electrophoresis with laser-induced fluorescence (CE-LIF) with conventional techniques.
Forensic science international (1998-06-17)
M Frost, H Köhler
ABSTRAKT
The increasing use of the potent hallucinogenic drug D-lysergic acid diethylamide (LSD) makes the application of highly sensitive analytical techniques necessary. For an effective analysis of biological specimens a limit of detection in the sub-ng/ml range is required. Using capillary electrophoresis (CE), with laser-induced fluorescence detection it was possible to determine 0.1-0.2 ng LSD/ml blood. The conventionally used immunoassays and gas and high-performance liquid chromatographic methods have cut-off values and limit of detection, respectively, in the same range. Only tandem mass spectrometry techniques can be more sensitive. Therefore, CE can be considered a good complementary method to conventional immunological and chromatographic techniques for the forensic analysis of biofluids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lysergic acid diethylamide