Merck
  • Synthesis and characterization of a novel copper (II)? silver (I) mixed-metal coordination polymer: Ag [Cu (2-pyrazinecarboxylate)< sub> 2</sub>](H< sub> 2</sub> O)(NO< sub> 3</sub>).
Synthesis and characterization of a novel copper (II)? silver (I) mixed-metal coordination polymer: Ag [Cu (2-pyrazinecarboxylate)&lt; sub&gt; 2&lt;/sub&gt;](H&lt; sub&gt; 2&lt;/sub&gt; O)(NO&lt; sub&gt; 3&lt;/sub&gt;).
Solid State Sciences (2000)
Dong Y-B, et al.