Merck
Metabolism of bisphenol A by hyper lignin-degrading fungus Phanerochaete sordida YK-624 under non-ligninolytic condition.
Chemosphere (2014-03-04)
Jianqiao Wang, Yuto Yamada, Akira Notake, Yasushi Todoroki, Toshinobu Tokumoto, Jing Dong, Peter Thomas, Hirofumi Hirai, Hirokazu Kawagishi
ABSTRAKT
Recently, we reported the conversion of bisphenol A (BPA) to 4-(2-(4-hydroxyphenyl)propan-2-yl)benzene-1,2-diol (hydroxy-BPA) by hyper lignin-degrading fungus Phanerochaete sordida YK-624 under non-ligninolytic condition. In the present study, the metabolism of hydroxy-BPA by P. sordida YK-624 was demonstrated under non-ligninolytic condition. Under these conditions, approximately 66% of hydroxy-BPA was degraded after 7 d of incubation. High-resolution electrospray ionization mass spectra and nuclear magnetic resonance analyses of the metabolites isolated from the culture broth indicated that hydroxy-BPA was metabolized to 4-(2-(4-hydroxyphenyl)propan-2-yl)-2-methoxyphenol (methoxy-BPA) and to 4-(2-(3,4-dimethoxyphenyl)propan-2-yl)phenol (dimethoxy-BPA) by sequential methylation events. These metabolites showed reduced estrogenic activity compared to BPA. These results suggested that the hydroxy BPA is methylated to two low toxic-methylation metabolites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bisphenol A, ≥99%