Merck
  • Limb remote ischemic preconditioning attenuates lung injury after pulmonary resection under propofol-remifentanil anesthesia: a randomized controlled study.
Limb remote ischemic preconditioning attenuates lung injury after pulmonary resection under propofol-remifentanil anesthesia: a randomized controlled study.
Anesthesiology (2014-04-20)
Cai Li, Miao Xu, Yan Wu, Yun-Sheng Li, Wen-Qi Huang, Ke-Xuan Liu
ABSTRAKT
Remote ischemic preconditioning (RIPC) may confer the protection in critical organs. The authors hypothesized that limb RIPC would reduce lung injury in patients undergoing pulmonary resection. In a randomized, prospective, parallel, controlled trial, 216 patients undergoing elective thoracic pulmonary resection under one-lung ventilation with propofol-remifentanil anesthesia were randomized 1:1 to receive either limb RIPC or conventional lung resection (control). Three cycles of 5-min ischemia/5-min reperfusion induced by a blood pressure cuff served as RIPC stimulus. The primary outcome was PaO2/FIO2. Secondary outcomes included other pulmonary variables, the incidence of in-hospital complications, markers of oxidative stress, and inflammatory response. Limb RIPC significantly increased PaO2/FIO2 compared with control at 30 and 60 min after one-lung ventilation, 30 min after re-expansion, and 6 h after operation (238 ± 52 vs. 192 ± 67, P = 0.03; 223 ± 66 vs. 184 ± 64, P = 0.01; 385 ± 61 vs. 320 ± 79, P = 0.003; 388 ± 52 vs. 317 ± 46, P = 0.001, respectively). In comparison with control, it also significantly reduced serum levels of interleukin-6 and tumor necrosis factor-α at 6, 12, 24, and 48 h after operation and malondialdehyde levels at 60 min after one-lung ventilation and 30 min after re-expansion (all P < 0.01). The incidence of acute lung injury and the length of postoperative hospital stay were markedly reduced by limb RIPC compared with control (all P < 0.05). Limb RIPC attenuates acute lung injury via improving intraoperative pulmonary oxygenation in patients without severe pulmonary disease after lung resection under propofol-remifentanil anesthesia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Propofol, European Pharmacopoeia (EP) Reference Standard
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2,6-Diisopropylphenol, 97%