Merck
Reversible protection of Cys-93(β) by PEG alters the structural and functional properties of the PEGylated hemoglobin.
Biochimica et biophysica acta (2014-04-22)
Qingqing Wang, Lijing Sun, Shaoyang Ji, Dawei Zhao, Jiaxin Liu, Zhiguo Su, Tao Hu
ABSTRAKT
As a potential hemoglobin (Hb)-based oxygen carrier (HBOC), the PEGylated Hb has received much attention for its non-nephrotoxicity. However, PEGylation can adversely alter the structural and functional properties of Hb. The site of PEGylation is an important factor to determine the structure and function of the PEGylated Hb. Thus, protection of some sensitive residues of Hb from PEGylation is of great significance to develop the PEGylated Hb as HBOC. Here, Cys-93(β) of Hb was conjugated with 20 kDa polyethylene glycol (PEG20K) through hydrazone and disulfide bonds. Then, the conjugate was modified with PEG5K succinimidyl carbonate (PEG5K-SC) using acylation chemistry, followed by removal of PEG20K Hb with hydrazone hydrolysis and disulfide reduction. Reversible conjugation of PEG20K at Cys-93(β) can protect Lys-95(β), Val-1(α) and Lys-16(α) of Hb from PEGylation with PEG5K-SC. The autoxidation rate, oxygen affinity, structural perturbation and tetramer instability of the PEGylated Hb were significantly decreased upon protection with PEG20K. The present study is expected to improve the efficacy of the PEGylated Hb as an oxygen therapeutic.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland