Merck
  • Proposed actions for the US Food and Drug Administration to implement to minimize adverse effects associated with energy drink consumption.
Proposed actions for the US Food and Drug Administration to implement to minimize adverse effects associated with energy drink consumption.
American journal of public health (2014-05-17)
Janet Thorlton, David A Colby, Paige Devine
ABSTRAKT
Energy drink sales are expected to reach $52 billion by 2016. These products, often sold as dietary supplements, typically contain stimulants. The Dietary Supplement Protection Act claims an exemplary public health safety record. However, in 2011 the number of emergency department visits related to consumption of energy drinks exceeded 20,000. Nearly half of these visits involved adverse effects occurring from product misuse. Political, social, economic, practical, and legal factors shape the landscape surrounding this issue. In this policy analysis, we examine 3 options: capping energy drink caffeine levels, creating a public education campaign, and increasing regulatory scrutiny regarding the manufacture and labeling of energy drinks. Increased regulatory scrutiny may be in order, especially in light of wrongful death lawsuits related to caffeine toxicity resulting from energy drink consumption.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Standardowa temperatura topnienia kofeiny, United States Pharmacopeia (USP) Reference Standard