Merck
Erythropoietin therapy after allogeneic hematopoietic cell transplantation: a prospective, randomized trial.
Blood (2014-05-23)
Aurélie Jaspers, Frédéric Baron, Evelyne Willems, Laurence Seidel, Kaoutar Hafraoui, Gaetan Vanstraelen, Christophe Bonnet, Yves Beguin
ABSTRAKT
We conducted a prospective randomized trial to assess hemoglobin (Hb) response to recombinant human erythropoietin (rhEPO) therapy after hematopoietic cell transplantation (HCT). Patients (N = 131) were randomized (1:1) between no treatment (control arm) or erythropoietin at 500 U/kg per week (EPO arm). Patients were also stratified into 3 cohorts: patients undergoing myeloablative HCT with rhEPO to start on day (D)28, patients given nonmyeloablative HCT (NMHCT) with rhEPO to start on D28, and patients also given NMHCT but with rhEPO to start on D0. The proportion of complete correctors (ie, Hb ≥13 g/dL) before D126 posttransplant was 8.1% in the control arm (median not reached) and 63.1% in the EPO arm (median, 90 days) (P < .001). Hb levels were higher and transfusion requirements decreased (P < .001) in the EPO arm, but not during the first month in the nonmyeloablative cohort starting rhEPO on D0. There was no difference in rates of thromboembolic events or other complications between the 2 arms. This is the first randomized trial to demonstrate that rhEPO therapy hastens erythroid recovery and decreases transfusion requirements when started one month after allogeneic HCT. There was no benefit to start rhEPO earlier after NMHCT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Erythropoietin, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture