Merck
Characterization of carbohydrates in rainwater from the southeastern North Carolina.
Chemosphere (2014-05-31)
Katherine M Mullaugh, Jade N Byrd, G Brooks Avery, Ralph N Mead, Joan D Willey, Robert J Kieber
ABSTRAKT
Carbohydrates have been widely reported in atmospheric aerosols, but have not previously been quantified in rainwater. We have identified and quantified a series of 11 specific compounds including monosaccharides (glucose, fructose, arabinose, galactose and pinitol), disaccharides (sucrose and trehalose), sugar alcohols (arabitol, dulcitol and mannitol) and the anhydrosaccharide levoglucosan. Rainwater analyzed in this study includes 52 distinct precipitation events in Wilmington, NC between June 2011 and October 2012. Our analysis indicates carbohydrates typically contribute <1% of total dissolved organic carbon in rain, but can account for as much as 10-35% during periods of high pollen or local fires. Concentrations of individual carbohydrates reached as high as 5.8 μM, with glucose and sucrose typically being the predominant species. The distribution of carbohydrates exhibited a distinct seasonal pattern, with higher concentrations of most carbohydrates, especially sucrose, in spring and summer, driven primarily by increased biogenic inputs during the growing season. Concentrations of carbohydrates were an order of magnitude higher in storms of terrestrial origin compared to marine events, further supporting a terrestrial biogenic origin of most species. Sequential sampling of Hurricane Irene showed significant quantities of carbohydrates present at the end of the storm when air mass back trajectories traversed over land. The highest level of levoglucosan, a compound associated with biomass burning, was detected in rain with an air mass back trajectory that traveled over a region affected by wildfires. When compared to aerosol concentrations reported by others, the sugar concentrations in rain demonstrate wet deposition is an important removal mechanism of this water-soluble and bioavailable fraction of atmospheric particulate organic matter.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Węgiel aktywowany, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Węgiel - Szklisty, rod, 200mm, diameter 1.0mm, glassy carbon
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis