Merck
Frontal and thalamic changes of GABA concentration indicate dysfunction of thalamofrontal networks in juvenile myoclonic epilepsy.
Epilepsia (2014-06-07)
Elke Hattingen, Christian Lückerath, Stefanie Pellikan, Dmitri Vronski, Christine Roth, Susanne Knake, Matthias Kieslich, Ulrich Pilatus
ABSTRAKT
Juvenile myoclonic epilepsy (JME) has been considered to be a frontal variant of thalamocortical network dysfunction in epilepsy. Changes of γ-aminobutyric acid (GABA)ergic neurotransmission may play a key role in this dysfunction. Magnetic resonance spectroscopy (MRS) is the only noninvasive method to measure GABA concentrations in different brain regions. We measured GABA and other metabolite concentrations in the thalamus and frontal lobe of patients with JME. A specific protocol was used for determining GABA concentrations in the thalamus, frontal lobe, and motor cortex contralateral to the handedness in 15 patients with JME and 15 age-matched controls. In addition, we measured concentrations of glutamate and glutamine, N-acetyl-aspartate (NAA), myoinositol, creatine, and choline using MRS with short echo time. JME-related concentration changes were analyzed comparing patients to controls, also considering potential effects of antiepileptic drugs. In patients with JME, GABA and NAA were reduced in the thalamus (p = 0.03 and p = 0.02), whereas frontal GABA and glutamine were elevated (p = 0.046 and p = 0.03). MRS revealed reduced NAA in the thalamic gray matter contralateral to the handedness (p = 0.04 each). These changes were found consistently in patients treated with new antiepileptic drugs and with valproate, although the extent of metabolic changes differed between these treatments. Decreased thalamic and increased frontal GABA suggest a dysfunction of GABAergic neurotransmission in these brain regions of patients with JME. The NAA decrease in the gray matter of the thalamus may hint to a damage of GABAergic neurons, whereas frontal increase of GABA and its precursor glutamine may reflect increased density in GABAergic neurons due to subtle cortical disorganization in the thalamofrontal network.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard