Merck
Beyond erythropoiesis: emerging metabolic roles of erythropoietin.
Diabetes (2014-06-26)
Minna Woo, Meredith Hawkins
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture
Erythropoietin, BRP, European Pharmacopoeia (EP) Reference Standard