Merck
The elemental composition of virus particles: implications for marine biogeochemical cycles.
Nature reviews. Microbiology (2014-06-17)
Luis F Jover, T Chad Effler, Alison Buchan, Steven W Wilhelm, Joshua S Weitz
ABSTRAKT
In marine environments, virus-mediated lysis of host cells leads to the release of cellular carbon and nutrients and is hypothesized to be a major driver of carbon recycling on a global scale. However, efforts to characterize the effects of viruses on nutrient cycles have overlooked the geochemical potential of the virus particles themselves, particularly with respect to their phosphorus content. In this Analysis article, we use a biophysical scaling model of intact virus particles that has been validated using sequence and structural information to quantify differences in the elemental stoichiometry of marine viruses compared with their microbial hosts. By extrapolating particle-scale estimates to the ecosystem scale, we propose that, under certain circumstances, marine virus populations could make an important contribution to the reservoir and cycling of oceanic phosphorus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%
Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 25x25mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 2mm, diameter 0.5mm, graphite, 99.95%
Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 200x200mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 25mm, graphite, 100%