Merck
Clostridium difficile infection in patients with ileal pouches.
The American journal of gastroenterology (2014-07-06)
Darren N Seril, Bo Shen
ABSTRAKT
Clostridium difficile (C. difficile) infection (CDI) following total proctocolectomy and ileal pouch-anal anastomosis has been increasingly recognized over the past 5 years. CDI of the ileal pouch has been recognized in ∼10% of symptomatic patients seen at a tertiary referral center for pouch dysfunction. In contrast to colonic CDI in the general population or in patients with inflammatory bowel disease, postoperative antibiotic exposure and the use of immunosuppressive agents or proton pump inhibitors do not appear to be associated with CDI of the pouch. Male gender, recent hospitalization, and presurgery antibiotic use were shown to be risk factors for ileal pouch CDI. The ileal pouch may be susceptible to CDI owing to similarities with the colon at physiological and structural levels. Postcolectomy CDI likely represents a spectrum of disease processes, varying from asymptomatic colonization to severe symptomatic infection. CDI should be considered in any patient with an ileal pouch presenting with a change in "normal" symptom pattern or treatment-refractory disease. Sensitive and specific methods for the detection of CDI are available, and pouchoscopy is a valuable tool in the evaluation of the patient with symptomatic CDI of the pouch. At a referral center for pouch dysfunction, vancomycin is used as the first-line therapy for ileal pouch CDI. Fecal microbiota transplantation may find use in the management of severe or antibiotic refractory CDI-related pouchitis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Vancomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Millipore
Vancomycin supplement, suitable for microbiology
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)