Merck
  • Comparison of cast materials for the treatment of congenital idiopathic clubfoot using the Ponseti method: a prospective randomized controlled trial.
Comparison of cast materials for the treatment of congenital idiopathic clubfoot using the Ponseti method: a prospective randomized controlled trial.
Canadian journal of surgery. Journal canadien de chirurgie (2014-08-01)
Catherine Hui, Elaine Joughin, Alberto Nettel-Aguirre, Simon Goldstein, James Harder, Gerhard Kiefer, David Parsons, Carmen Brauer, Jason Howard
ABSTRAKT
The Ponseti method of congenital idiopathic clubfoot correction has traditionally specified plaster of Paris (POP) as the cast material of choice; however, there are negative aspects to using POP. We sought to determine the influence of cast material (POP v. semirigid fibreglass [SRF]) on clubfoot correction using the Ponseti method. Patients were randomized to POP or SRF before undergoing the Ponseti method. The primary outcome measure was the number of casts required for clubfoot correction. Secondary outcome measures included the number of casts by severity, ease of cast removal, need for Achilles tenotomy, brace compliance, deformity relapse, need for repeat casting and need for ancillary surgical procedures. We enrolled 30 patients: 12 randomized to POP and 18 to SRF. There was no difference in the number of casts required for clubfoot correction between the groups (p = 0.13). According to parents, removal of POP was more difficult (p < 0.001), more time consuming (p < 0.001) and required more than 1 method (p < 0.001). At a final follow-up of 30.8 months, the mean times to deformity relapse requiring repeat casting, surgery or both were 18.7 and 16.4 months for the SRF and POP groups, respectively. There was no significant difference in the number of casts required for correction of clubfoot between the 2 materials, but SRF resulted in a more favourable parental experience, which cannot be ignored as it may have a positive impact on psychological well-being despite the increased cost associated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%