Merck
Reply: The importance of genotype variation beyond different antiplatelet therapy in nonresponder patients.
Journal of the American College of Cardiology (2014-07-19)
Dániel Aradi, András Komócsi
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard