Merck
Schedules of controlled substances: rescheduling of hydrocodone combination products from schedule III to schedule II. Final rule.
Federal register (2014-08-30)
ABSTRAKT
With the issuance of this final rule, the Administrator of the Drug Enforcement Administration reschedules hydrocodone combination products from schedule III to schedule II of the Controlled Substances Act. This scheduling action is pursuant to the Controlled Substances Act which requires that such actions be made on the record after opportunity for a hearing through formal rulemaking. This action imposes the regulatory controls and administrative, civil, and criminal sanctions applicable to schedule II controlled substances on persons who handle (manufacture, distribute, dispense, import, export, engage in research, conduct instructional activities with, conduct chemical analysis with, or possess) or propose to handle hydrocodone combination products.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrocodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®