Merck
Crystal structure of the human T cell receptor CD3 epsilon gamma heterodimer complexed to the therapeutic mAb OKT3.
Proceedings of the National Academy of Sciences of the United States of America (2004-05-12)
Lars Kjer-Nielsen, Michelle A Dunstone, Lyudmila Kostenko, Lauren K Ely, Travis Beddoe, Nicole A Mifsud, Anthony W Purcell, Andrew G Brooks, James McCluskey, Jamie Rossjohn
ABSTRAKT
The CD3 epsilon gamma heterodimer is essential for expression and function of the T cell receptor. The crystal structure of the human CD3 epsilon gamma heterodimer is described to 2.1-A resolution complexed with OKT3, a therapeutic mAb that not only activates and tolerizes mature T cells but also induces regulatory T cells. The mode of CD3 epsilon gamma dimerization provides a general structural basis for CD3 assembly and maps candidate T cell antigen receptor docking sites, including a duplicated linear region rich in acidic residues that is unique to human CD3 epsilon. OKT3 binds to an atypically small area of CD3 epsilon and has a low affinity for the isolated CD3 epsilon gamma heterodimer. The structure of the OKT3/CD3 epsilon gamma complex has implications for T cell signaling and therapeutic design.