Merck
Surface and biocidal activity of some synthesized metallo azobenzene isothiouronium salts.
Bioorganic & medicinal chemistry (2006-09-16)
A M Badawi, E M S Azzam, S M I Morsy
ABSTRAKT
A novel series of azobenzene isothiouronium salts of different alkyl chains (propyl, hexyl and dodecyl) were synthesized by reaction of 4-((4-methylphenyl)azo)phenol with 1,3-dibromopropane, 1,6-dibromohexane and 1,12-dibromododecane, respectively. These salts were reacted with copper (II) halide to give their corresponding metallo complexes. The surface tension measurements for the synthesized compounds show that the metallo complexes have adsorption and micellization better than that of the parent azobenzene isothiouronium salts. The pathogenic Gram-negative bacteria, Gram-positive bacteria, fungi and yeast were used to determine the biocidal activity of these compounds using gradient plate technique. The results indicate that the copper complexes of the synthesized azobenzene isothiouronium salts have a relatively better biocidal activity than the parent salts.