Merck
EDEM1 accelerates the trimming of alpha1,2-linked mannose on the C branch of N-glycans.
Glycobiology (2010-01-13)
Nobuko Hosokawa, Linda O Tremblay, Barry Sleno, Yukiko Kamiya, Ikuo Wada, Kazuhiro Nagata, Koichi Kato, Annette Herscovics
ABSTRAKT
Glycoprotein folding and degradation in the endoplasmic reticulum (ER) is mediated by the ER quality control system. Mannose trimming plays an important role by forming specific N-glycans that permit the recognition and sorting of terminally misfolded conformers for ERAD (ER-associated degradation). The EDEM (ER degradation enhancing alpha-mannosidase-like protein) subgroup of proteins belonging to the Class I alpha1,2-mannosidase family (glycosylhydrolase family 47) has been shown to enhance ERAD. We recently reported that overexpression of EDEM3 enhances glycoprotein ERAD with a concomitant increase in mannose-trimming activity in vivo. Herein, we report that overexpression of EDEM1 produces Glc(1)Man(8)GlcNAc(2) isomer C on terminally misfolded null Hong Kong alpha1-antitrypsin (NHK) in vivo. Levels of this isomer increased throughout the chase period and comprised approximately 10% of the [(3)H]mannose-labeled N-glycans on NHK after a 3-h chase. Furthermore, overexpression of EDEM1 E220Q containing a mutation in a conserved catalytic residue essential for alpha1,2-mannosidase activity did not yield detectable levels of Glc(1)Man(8)GlcNAc(2) isomer C. Yet, the same extent of NHK ERAD-enhancement was observed in both EDEM1 and EDEM1 E220Q overexpressing cells. This can be attributed to both wild-type and mutant EDEM1 inhibiting aberrant NHK dimer formation. We further analyzed the N-glycan profile of total cellular glycoproteins from HepG2 cells stably overexpressing EDEM1 and found that the relative amount of Man(7)GlcNAc(2) isomer A, which lacks the terminal B and C branch mannoses, was increased compared to parental HepG2 cells. Based on this observation, we conclude that EDEM1 activity trims mannose from the C branch of N-glycans in vivo.