Merck
Identification of a BRCA1-mRNA splicing complex required for efficient DNA repair and maintenance of genomic stability.
Molecular cell (2014-04-22)
Kienan I Savage, Julia J Gorski, Eliana M Barros, Gareth W Irwin, Lorenzo Manti, Alexander J Powell, Andrea Pellagatti, Natalia Lukashchuk, Dennis J McCance, W Glenn McCluggage, Giuseppe Schettino, Manuel Salto-Tellez, Jacqueline Boultwood, Derek J Richard, Simon S McDade, D Paul Harkin
ABSTRAKT
Mutations within BRCA1 predispose carriers to a high risk of breast and ovarian cancers. BRCA1 functions to maintain genomic stability through the assembly of multiple protein complexes involved in DNA repair, cell-cycle arrest, and transcriptional regulation. Here, we report the identification of a DNA damage-induced BRCA1 protein complex containing BCLAF1 and other key components of the mRNA-splicing machinery. In response to DNA damage, this complex regulates pre-mRNA splicing of a number of genes involved in DNA damage signaling and repair, thereby promoting the stability of these transcripts/proteins. Further, we show that abrogation of this complex results in sensitivity to DNA damage, defective DNA repair, and genomic instability. Interestingly, mutations in a number of proteins found within this complex have been identified in numerous cancer types. These data suggest that regulation of splicing by the BRCA1-mRNA splicing complex plays an important role in the cellular response to DNA damage.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.