Merck
Spectroscopic study on the photophysical properties of lanthanide complexes with 2, 2'-bipyridine-N, N'-dioxide.
Journal of fluorescence (2004-12-24)
Bing Yan, Yi Shan Song
ABSTRAKT
Five novel lanthanide (Eu3+, Tb3+, Gd3+, Sm3+ and Dy3+) complexes with 2, 2'-bipyridine-N, N'-dioxide (bipyO2) were synthesized and characterized by elemental analysis, IR spectrum. The triplet state energy of bipy O2 was determined to be 22275 cm(-1) with the phosphorescence spectrum of bipy O2 and its gadolinium complex. The photophysical properties of these complexes indicated that the triplet state energy of the ligand is suitable for the sensitization of the luminescence of Eu3+, Tb3+, Sm3+ and Dy3+, especially of Tb3+.