- Clean synthesis of triarylamines: Buchwald-Hartwig reaction in water with amphiphilic resin-supported palladium complexes.
Clean synthesis of triarylamines: Buchwald-Hartwig reaction in water with amphiphilic resin-supported palladium complexes.
Chemical communications (Cambridge, England) (2010-02-04)
Yoshinori Hirai, Yasuhiro Uozumi
PMID20126727
ABSTRAKT
Catalytic aromatic amination was achieved in water under heterogeneous conditions by the use of palladium complexes anchored to the amphiphilic PS-PEG resin with little palladium leaching to provide a green and clean (metal-uncontaminated) protocol for the preparation of triarylamines, including the optoelectronically active N,N,N',N'-tetraaryl-1,1'-biphenyl-4,4'-diamines (TPDs).