- (E)-2-[4-(Trifluoro-meth-oxy)benzyl-idene]indan-1-one.
(E)-2-[4-(Trifluoro-meth-oxy)benzyl-idene]indan-1-one.
Acta crystallographica. Section E, Structure reports online (2011-11-18)
Mohamed Ashraf Ali, Rusli Ismail, Soo Choon Tan, Mohd Mustaqim Rosli, Hoong-Kun Fun
PMID22091162
ABSTRAKT
In the title compound, C(17)H(11)F(3)O(2), the dihydro-indene ring is approximately planar with a maximum deviation of 0.024 (2) Å and makes a dihedral angle of 3.17 (8) Å with the adjacent benzene ring. In the crystal, mol-ecules are inter-connected by C-H⋯O inter-actions, forming an infinite chain along the c axis.