Merck
  • Aerosol interleukin-2 induces natural killer cell proliferation in the lung and combination therapy improves the survival of mice with osteosarcoma lung metastasis.
Aerosol interleukin-2 induces natural killer cell proliferation in the lung and combination therapy improves the survival of mice with osteosarcoma lung metastasis.
Pediatric blood & cancer (2014-03-13)
Sergei R Guma, Dean A Lee, Yu Ling, Nancy Gordon, Eugenie S Kleinerman
ABSTRAKT
We have previously shown that aerosol interleukin-2 (IL-2) increased the number of intravenously injected human natural killer (NK) cells in the lungs. In this study we investigated whether this increase was secondary to NK cell proliferation and determined the site of the proliferation. Nude mice with osteosarcoma lung metastases were injected with NK cells and treated with aerosol IL-2 or aerosol PBS. BrdU was injected prior to euthanasia to identify proliferating NK cells. The percentage of proliferating NK cells in the lung, bone marrow, spleen, and liver was determined using flow cytometry. Survival studies for mice with osteosarcoma lung metastasis treated with aerosol PBS, aerosol IL-2 alone, aerosol PBS plus NK cells, and aerosol IL-2 plus NK cells were also performed. Treatment with aerosol IL-2 induced the proliferation of injected NK cells in the lung. Aerosol IL-2 did not increase the proliferation of NK cells in the spleen and liver. Treatment with aerosol IL-2 and aerosol IL-2 plus NK cells increased the overall survival of mice with osteosarcoma lung metastasis. Aerosol IL-2 increases lung NK cell numbers by stimulating local NK cell proliferation. Aerosol IL-2's effect on NK cell proliferation is organ specific, which makes it ideal for the specific targeting of lung metastasis. Aerosol IL-2 plus NK cell therapy induced metastatic regression and increased overall survival demonstrating the potential of this therapeutic approach for patients with osteosarcoma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Supelco
Roztwór jonów amonowych dla ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%