Merck
Combined effects of lanthanum (III) chloride and acid rain on photosynthetic parameters in rice.
Chemosphere (2014-07-23)
Lihong Wang, Wen Wang, Qing Zhou, Xiaohua Huang
ABSTRAKT
Rare earth elements (REEs) pollution and acid rain are environmental issues, and their deleterious effects on plants attract worldwide attention. These two issues exist simultaneously in many regions, especially in some rice-growing areas. However, little is known about the combined effects of REEs and acid rain on plants. Here, the combined effects of lanthanum chloride (LaCl3), one type of REE salt, and acid rain on photosynthesis in rice were investigated. We showed that the combined treatment of 81.6 μM LaCl3 and acid rain at pH 4.5 increased net photosynthetic rate (Pn), stomatic conductance (Gs), intercellular CO2 concentration (Ci), Hill reaction activity (HRA), apparent quantum yield (AQY) and carboxylation efficiency (CE) in rice. The combined treatment of 81.6 μM LaCl3 and acid rain at pH 3.5 began to behave toxic effects on photosynthesis (decreasing Pn, Gs, HRA, AQY and CE, and increasing Ci), and the maximally toxic effects were observed in the combined treatment of 2449.0 μM LaCl3 and acid rain at pH 2.5. Moreover, the combined effects of LaCl3 and acid rain on photosynthesis in rice depended on the growth stage of rice, with the maximal effects occurring at the booting stage. Furthermore, the combined treatment of high-concentration LaCl3 and low-pH acid rain had more serious effects on photosynthesis in rice than LaCl3 or acid rain treatment alone. Finally, the combined effect of LaCl3 and acid rain on Pn in rice resulted from the changes in stomatic (Gs, Ci) and non-stomatic (HRA, AQY and CE) factors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, ACS reagent
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%