Merck
Bacterial acquisition of hexachlorobenzene-derived carbon in contaminated soil.
Chemosphere (2014-07-30)
Ondrej Uhlik, Michal Strejcek, Jan Vondracek, Lucie Musilova, Jakub Ridl, Petra Lovecka, Tomas Macek
ABSTRAKT
Pesticides are a class of xenobiotics intentionally released into the environment. Hexachlorobenzene (HCB) was used as a fungicide from 1945, leaving behind many contaminated sites. Very few studies have examined the biodegradation of HCB or the fate of HCB-derived carbon. Here we report that certain bacterial populations are capable of deriving carbon from HCB in contaminated soil under aerobic conditions. These populations are primarily Proteobacteria, including Methylobacterium and Pseudomonas, which predominated as detected by stable isotope probing (SIP) and 16S rRNA gene amplicon pyrosequencing. Due to the nature of SIP, which can be used as a functional method solely for assimilatory processes, it is not possible to elucidate whether these populations metabolized directly HCB or intermediates of its metabolism produced by different populations. The possibility exists that HCB is degraded via the formation of pentachlorophenol (PCP), which is further mineralized. With this in mind, we designed primers to amplify PCP 4-monooxygenase-coding sequences based on the available pcpB gene sequence from Methylobacterium radiotolerans JCM 2831. Based on 16S rRNA gene analysis, organisms closely related to this strain were detected in (13)C-labeled DNA. Using the designed primers, we were able to amplify pcpB genes in both total community DNA and (13)C-DNA. This indicates that HCB might be transformed into PCP before it gets assimilated. In summary, this study is the first report on which bacterial populations benefit from carbon originating in the pesticide HCB in a contaminated soil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Pentachlorophenol solution, certified reference material, 5000 μg/mL in methanol
Supelco
Hexachlorobenzene solution, certified reference material, 1000 μg/mL in acetone
Sigma-Aldrich
Pentachlorophenol, 97%
Supelco
Pentachlorophenol, analytical standard
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard