Merck
  • An increasing opine carbon bias in artificial exudation systems and genetically modified plant rhizospheres leads to an increasing reshaping of bacterial populations.
An increasing opine carbon bias in artificial exudation systems and genetically modified plant rhizospheres leads to an increasing reshaping of bacterial populations.
Molecular ecology (2014-08-26)
Samuel Mondy, Aurore Lenglet, Amelie Beury-Cirou, Celestin Libanga, Pascal Ratet, Denis Faure, Yves Dessaux
ABSTRAKT
To investigate how exudation shapes root-associated bacterial populations, transgenic Arabidopsis thaliana plants that exuded the xenotopic compound octopine at low and high rates were grown in a nonsterile soil. Enumerations of both cultivable and octopine-degrading bacteria demonstrated that the ratios of octopine degraders increased along with octopine concentration. An artificial exudation system was also set up in which octopine was brought at four ratios. The density of octopine-degrading bacteria directly correlated with the input of octopine. Bacterial diversity was analysed by rrs amplicon pyrosequencing. Ensifer and Pseudomonas were significantly more frequently detected in soil amended with artificial exudates. However, the density of Pseudomonas increased as a response to carbon supplementation while that of Ensifer only correlated with octopine concentrations possibly in relation to two opposed colonization strategies of rhizosphere bacteria, that is, copiotrophy and oligotrophy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Carbon, microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, microleaf, 25x70mm, thinness 0.050μm, specific density 10μg/cm2, temporary glass support, 99.997%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%
Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.25μm, specific density 50μg/cm2, temporary glass support, 99.997%
Carbon, foil, 25x25mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 2mm, diameter 0.5mm, graphite, 99.95%
Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 200x200mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 2.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 25mm, graphite, 100%