Merck
Glyceraldehyde-3-phosphate dehydrogenase gene over expression correlates with poor prognosis in non small cell lung cancer patients.
Molecular cancer (2013-08-31)
Roberto Puzone, Graziana Savarino, Sandra Salvi, Maria Giovanna Dal Bello, Giulia Barletta, Carlo Genova, Erika Rijavec, Claudio Sini, Alessia Isabella Esposito, Giovanni Battista Ratto, Mauro Truini, Francesco Grossi, Ulrich Pfeffer
ABSTRAKT
Glycolysis in presence of oxygen with high glucose consumption is known to be the metabolism of choice in many tumors. In lung cancer this phenomenon is routinely exploited in diagnostic PET imaging of fluorodeoxyglucose uptake, but not much is known about the prognostic capabilities of glycolysis level assessment in resected lung tumor samples. In this retrospective study, we used real time polymerase chain reaction(RQ-PCR) to assess the expression level of the gene for Glyceraldehyde 3-phosphate dehydrogenase(GAPDH), key enzyme for glucose breakdown, in tumor samples from 82 consecutive early stages resected non small cell lung cancer(NSCLC) patients. We then compared our results in six large publicly available NSCLC microarray datasets collecting data from over 1250 total patients. In our study GAPDH gene over expression was found to be an adverse prognostic factor in early stages NSCLC (n = 82 HR = 1.30 p = 0.050). This result was confirmed in 5 of 6 public datasets analyzed: Shedden et al. 2008: n = 442 HR = 1.54 p < 0.0001; Lee et al. 2008: n = 138 HR = 1.31 p = 0.043; Tomida et al. 2009: n = 117 HR = 1.59 p = 0.004; Roepman et al. 2009: n = 172 (TPI1 gene) HR = 1.51 p = 0.009; Okayama et al. 2012: n = 226 HR = 3.19 p < 0.0001; Botling et al. 2013: n = 196 HR = 1.00 p = 0.97). Furthermore, in the large and clinically well annotated Shedden et al. microarray dataset, GAPDH hazard ratio did not change whether calculated for the whole dataset or for the subgroup of adjuvant naive patients only (n = 330 HR = 1.49 p < 0.0001). GAPDH gene over expression in resected tumor samples is an adverse prognostic factor in NSCLC. Our results confirm the prognostic value of glucose metabolism assessment in NSCLC.