Merck
TULIP1 (RALGAPA1) haploinsufficiency with brain development delay.
Genomics (2009-09-08)
Keiko Shimojima, Yuta Komoike, Jun Tohyama, Sonoko Takahashi, Marco T Páez, Eiji Nakagawa, Yuichi Goto, Kousaku Ohno, Mayu Ohtsu, Hirokazu Oguni, Makiko Osawa, Toru Higashinakagawa, Toshiyuki Yamamoto
ABSTRAKT
A novel microdeletion of 14q13.1q13.3 was identified in a patient with developmental delay and intractable epilepsy. The 2.2-Mb deletion included 15 genes, of which TULIP1 (approved gene symbol: RALGAPA1)was the only gene highly expressed in the brain. Western blotting revealed reduced amount of TULIP1 in cell lysates derived from immortalized lymphocytes of the patient, suggesting the association between TULIP1 haploinsufficiency and the patient's phenotype, then 140 patients were screened for TULIP1 mutations and four missense mutations were identified. Although all four missense mutations were common with parents, reduced TULIP1 was observed in the cell lysates with a P297T mutation identified in a conserved region among species. A full-length homolog of human TULIP1 was identified in zebrafish with 72% identity to human. Tulip1 was highly expressed in zebrafish brain, and knockdown of which resulted in brain developmental delay. Therefore, we suggest that TULIP1 is a candidate gene for developmental delay.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid