Merck
Analysis of gene promoter regulation by tumor suppressor genes.
Methods in molecular biology (Clifton, N.J.) (2003-06-05)
Kumaravel Somasundaram, Sanjeev Das, Smita Lakhotia, Narendra Wajapeyee
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Coenzyme A sodium salt hydrate