Merck
  • Association of protein expression in isolated milk epithelial cells and cis-9, trans-11 conjugated linoleic acid proportions in milk from dairy cows.
Association of protein expression in isolated milk epithelial cells and cis-9, trans-11 conjugated linoleic acid proportions in milk from dairy cows.
Journal of the Science of Food and Agriculture (2014)
Wang T, Lim JN, Bok JD, et al.