Merck
Role of nucleus accumbens shell neuronal ensembles in context-induced reinstatement of cocaine-seeking.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-05-30)
Fabio C Cruz, Klil R Babin, Rodrigo M Leao, Evan M Goldart, Jennifer M Bossert, Yavin Shaham, Bruce T Hope
ABSTRAKT
Environmental contexts previously associated with drug use provoke relapse to drug use in humans and reinstatement of drug seeking in animal models of drug relapse. We examined whether context-induced reinstatement of cocaine seeking is mediated by activation of context-selected nucleus accumbens neurons. We trained rats to self-administer cocaine in Context A and extinguished their lever-pressing in a distinct Context B. On test day, reexposure to the cocaine-associated Context A reinstated cocaine seeking and increased expression of the neural activity marker Fos in 3.3% of accumbens shell and 1.6% of accumbens core neurons. To assess a causal role for these activated neurons, we used the Daun02 inactivation procedure to selectively inactivate these neurons. We trained c-fos-lacZ transgenic rats to self-administer cocaine in Context A and extinguished their lever-pressing in Context B. On induction day, we exposed rats to either Context A or a novel Context C for 30 min and injected Daun02 or vehicle into accumbens shell or core 60 min later. On test day, 3 d after induction day, the ability of Context A to reinstate cocaine seeking and increase neuronal activity in accumbens shell was attenuated when Daun02 was previously injected after exposure to Context A. Daun02 injections after exposure to the novel Context C had no effect on context-induced reinstatement of cocaine seeking despite much greater numbers of Fos-expressing neurons induced by Context C. Daun02 injections in accumbens core had no effect. Our data suggest that context-induced reinstatement of cocaine seeking is mediated by activation of context-selected accumbens shell but not core neuronal ensembles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat