Merck
Cytochemical localization of H2O2 in biological tissues.
Methods in molecular biology (Clifton, N.J.) (2002-08-03)
E Ann Ellis, Maria B Grant
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥94% (HPLC)
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor