Merck
Isolation and long-term cell culture of epithelial-like cells from rat liver.
Experimental cell research (1971-11-01)
G M Williams, E K Weisburger, J H Weisburger
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Williams′ Medium E, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Williams′ Medium E, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture