Merck
Immunohistochemistry of markers of histiomonocytic cells in malignant fibrous histiocytomas. A monoclonal antibody study.
Pathology, research and practice (1990-12-01)
Y Soini, M Miettinen
ABSTRAKT
Nine cases of malignant fibrous histiocytomas (MFH) were examined immunohistochemically in frozen sections with six different monoclonal antibodies to histiomonocytic and related cells (EBM11, HAM-56, KB90, antibodies to dendritic reticulum cells, HLADR and LCA). Ten other soft tissue sarcomas, two desmoid tumors, twelve carcinomas, three seminomas and four lymphomas were studied for comparison. All cases of MFH showed positivity for histiomonocytic cell antigens. In six cases, the positive cells could be clearly interpreted to be infiltrating non-neoplastic cells. However, immunoreactivity for multiple histiocytic markers (EBM11, HAM-56, KB90, HLADR) was seen in tumor cells in three cases of MFH. In one of these cases, the positivity could be verified with KP1, an antibody to histiomonocytic cells applied in formalin fixed and paraffin embedded tissue. None of the tumors was positive with the antibody to dendritic reticulum cells or LCA. In the series of non-histiocytic tumors, no cases showed widespread positivity for multiple histiocytic markers. Our results suggest that in relation to true histiomonocytic differentiation MFH might be a heterogeneous group of tumors. The widespread immunoreactivity for multiple histiocytic markers in some cases may indicate a true histiomonocytic differentiation in some MFHs.