Merck
Propranolol for infantile haemangiomas: single centre experience of 250 cases and proposed therapeutic protocol.
Archives of disease in childhood (2014-08-16)
Lea Solman, Amera Murabit, Maria Gnarra, John I Harper, Samira B Syed, Mary Glover
ABSTRAKT
To assess the safety and efficacy of systemic propranolol for the treatment of complicated infantile haemangiomas. Retrospective review of case notes of paediatric patients treated with propranolol for complicated infantile haemangiomas. Tertiary care children's hospital. All paediatric patients with complicated infantile haemangiomas who commenced treatment with propranolol from July 2008 to December 2011 and have completed treatment for at least 3 months. 250 patients were treated with propranolol; 34.4% were premature and 5.6% postmature. Indications for propranolol included: vision compromise (42.0%), bleeding and/or ulceration (30.4%) airway obstruction (8.8%), feeding difficulty (8.4%), risk of permanent disfigurement (4.4%) and other (6%) (nasal obstruction, auditory canal obstruction, large haemangioma, compression of neck structure and spinal cord). Median age at beginning of treatment was 4.5 months. Median age at end of treatment was 16.7 months. Median length of therapy was 11.8 months. Adverse effects (such as wheezing, worsening of ulceration, sleep disturbance, diarrhoea) occurred in 38 patients (15.2%), leading to modifications in management in 26 patients (10.4%). 240 patients (96%) had good to excellent response to treatment. 20 patients (8%) experienced regrowth of the haemangioma on cessation of propranolol and six patients (2.4%) required propranolol to be restarted. In appropriately selected patients, propranolol is a safe and effective treatment for infantile haemangiomas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Propranolol, United States Pharmacopeia (USP) Reference Standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
Propranolol hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-Propranolol hydrochloride, analytical standard