Merck
  • GSK2374697, a novel albumin-binding domain antibody (AlbudAb), extends systemic exposure of exendin-4: first study in humans--PK/PD and safety.
GSK2374697, a novel albumin-binding domain antibody (AlbudAb), extends systemic exposure of exendin-4: first study in humans--PK/PD and safety.
Clinical pharmacology and therapeutics (2014-09-23)
R L O'Connor-Semmes, J Lin, R J Hodge, S Andrews, J Chism, A Choudhury, D J Nunez
ABSTRAKT
GSK2374697 is a genetically engineered fusion protein of a human domain antibody to exendin-4. This molecule binds with a high affinity to human serum albumin, creating a long-duration glucagon-like peptide-1 (GLP-1) receptor agonist. This study is the first evaluation of the albumin-binding domain antibody (AlbudAb) drug delivery platform in humans. The aim of this randomized clinical study was to determine the pharmacokinetics, pharmacodynamics, safety, and tolerability of GSK2374697. The pharmacokinetic profile was prolonged, with estimated half-lives ranging from 6 to 10 days. Postprandial glucose and insulin were reduced, and gastric emptying was delayed in healthy subjects, confirming anticipated GLP-1 receptor agonist pharmacology. The safety and tolerability were as expected for a potent GLP-1 agonist. Gradual titration of doses greatly improved tolerability. Rapid tolerance to nausea was observed. Study results support further investigation in type 2 diabetes and for weight loss.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Paracetamol, European Pharmacopoeia (EP) Reference Standard
USP
Acetaminofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®