Merck
Parapedobacter indicus sp. nov., isolated from hexachlorocyclohexane-contaminated soil.
International journal of systematic and evolutionary microbiology (2014-10-10)
Roshan Kumar, Vatsala Dwivedi, Namita Nayyar, Helianthous Verma, Amit Kumar Singh, Pooja Rani, D L N Rao, Rup Lal
ABSTRAKT
Strain RK1(T), a Gram-stain-negative, non-spore-forming, rod-shaped, non-motile bacterium was isolated from a hexachlorocyclohexane (HCH) dumpsite, Lucknow, India. 16S rRNA gene sequence analysis revealed that strain RK1(T) belongs to the family Sphingobacteriaceae and showed highest sequence similarity to Parapedobacter koreensis Jip14(T) (95.63%). The major cellular fatty acids of strain RK1(T) were iso-C15:0, summed feature 3 (C16:1ω7c and/or C16:1ω6c), iso-C17:0 3-OH, summed feature 9 (10-methyl C16:0 and/or iso-C17:1ω9c), iso-C15:0 3-OH and C16 : 0. The major respiratory pigment and polyamine of RK1(T) were menaquinone (MK-7) and homospermidine, respectively. The main polar lipids were phosphatidylethanolamine and sphingolipid. The G+C content of the DNA was 44.5 mol%. The results of physiological and biochemical tests and 16S rRNA sequence analysis clearly demonstrated that strain RK1(T) represents a novel species of the genus Parapedobacter, for which the name Parapedobacter indicus sp. nov. is proposed. The type strain is RK1(T) ( = DSM 28470(T) =MCC 2546(T)).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Spermidyna 0,1 M
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Supelco
Spermidine, analytical standard
Supelco
Lindane, PESTANAL®, analytical standard
Supelco
Lindane, Pharmaceutical Secondary Standard; Certified Reference Material