Merck
Angiotensin blockade in late autosomal dominant polycystic kidney disease.
The New England journal of medicine (2014-11-18)
Vicente E Torres, Kaleab Z Abebe, Arlene B Chapman, Robert W Schrier, William E Braun, Theodore I Steinman, Franz T Winklhofer, Godela Brosnahan, Peter G Czarnecki, Marie C Hogan, Dana C Miskulin, Frederic F Rahbari-Oskoui, Jared J Grantham, Peter C Harris, Michael F Flessner, Charity G Moore, Ronald D Perrone
ABSTRAKT
Hypertension develops early in patients with autosomal dominant polycystic kidney disease (ADPKD) and is associated with disease progression. The renin-angiotensin-aldosterone system (RAAS) is implicated in the pathogenesis of hypertension in patients with ADPKD. Dual blockade of the RAAS may circumvent compensatory mechanisms that limit the efficacy of monotherapy with an angiotensin-converting-enzyme (ACE) inhibitor or angiotensin II-receptor blocker (ARB). In this double-blind, placebo-controlled trial, we randomly assigned 486 patients, 18 to 64 years of age, with ADPKD (estimated glomerular filtration rate [GFR], 25 to 60 ml per minute per 1.73 m(2) of body-surface area) to receive an ACE inhibitor (lisinopril) and placebo or lisinopril and an ARB (telmisartan), with the doses adjusted to achieve a blood pressure of 110/70 to 130/80 mm Hg. The composite primary outcome was the time to death, end-stage renal disease, or a 50% reduction from the baseline estimated GFR. Secondary outcomes included the rates of change in urinary aldosterone and albumin excretion, frequency of hospitalizations for any cause and for cardiovascular causes, incidence of pain, frequency of ADPKD-related symptoms, quality of life, and adverse study-medication effects. Patients were followed for 5 to 8 years. There was no significant difference between the study groups in the incidence of the composite primary outcome (hazard ratio with lisinopril-telmisartan, 1.08; 95% confidence interval, 0.82 to 1.42). The two treatments controlled blood pressure and lowered urinary aldosterone excretion similarly. The rates of decline in the estimated GFR, urinary albumin excretion, and other secondary outcomes and adverse events, including hyperkalemia and acute kidney injury, were also similar in the two groups. Monotherapy with an ACE inhibitor was associated with blood-pressure control in most patients with ADPKD and stage 3 chronic kidney disease. The addition of an ARB did not alter the decline in the estimated GFR. (Funded by the National Institute of Diabetes and Digestive and Kidney Diseases and others; HALT-PKD [Study B] ClinicalTrials.gov number, NCT01885559.).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Supelco
Aldosterone solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Telmisartan, European Pharmacopoeia (EP) Reference Standard
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard
USP
Telmisartan, United States Pharmacopeia (USP) Reference Standard
Lisinopril dihydrate, European Pharmacopoeia (EP) Reference Standard
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard